- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Olfr61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146964.1 | NP_667175.1 | 1 | 123 | ||
GenBank mRNA | BC119308.1 | - | 1 | 219 | 8734057 | |
BC119334.1 | - | 1 | 219 | 8734083 |