Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025378.2 | NP_001020549.1 | 9 | 857 | ||
NM_001271526.1 | NP_001258455.1 | 12 | 1382 | |||
NM_008765.3 | NP_032791.1 | 12 | 1388 | |||
GenBank mRNA | AB190256.1 | - | 9 | 847 | ||
AK048408.1 | - | 12 | 1207 | |||
AK051747.1 | - | 12 | 1217 | |||
AK133604.1 | - | 12 | 1208 | |||
AK146281.1 | - | 12 | 1207 | |||
AK146859.1 | - | 12 | 1202 | |||
BC015257.1 | - | 12 | 1198 | 4234871 | ||
U27457.1 | - | 12 | 1207 |