Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025570.1 | NP_001020741.1 | 2 | 1354 | ||
NM_011127.2 | NP_035257.1 | 2 | 1354 | |||
NM_175686.3 | NP_783617.2 | 2 | 1354 | |||
GenBank mRNA | AB221686.1 | - | 2 | 339 | ||
AK008428.2 | - | 3 | 184 | |||
AK019503.1 | - | 2 | 165 | |||
AK031075.1 | - | 2 | 320 | |||
AK032773.1 | - | 2 | 921 | |||
AK084387.1 | - | 2 | 1372 | |||
AK089006.1 | - | 2 | 1186 | |||
AK165914.1 | - | 2 | 127 | |||
BC092372.1 | - | 2 | 703 | 30533016 | ||
L06502.1 | - | 2 | 519 | |||
U03873.1 | - | 2 | 1360 | |||
X59725.1 | - | 2 | 480 |