Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou4f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011143.4 | NP_035273.3 | 1 | 331 | ||
GenBank mRNA | AY706205.1 | - | 1 | 331 |