Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293622.1 | NP_001280551.1 | 4 | 1251 | ||
NM_008913.5 | NP_032939.1 | 4 | 1251 | |||
GenBank mRNA | AK146387.1 | - | 4 | 1195 | ||
AK150393.1 | - | 4 | 610 | |||
AK166287.1 | - | 4 | 1028 | |||
BC138612.1 | - | 4 | 524 | 8861632 | ||
J05479.1 | - | 4 | 548 |