Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253890.1 | NP_001240819.1 | 2 | 189 | ||
NM_008923.3 | NP_032949.3 | 2 | 191 | |||
GenBank mRNA | AK158994.1 | - | 2 | 190 | ||
AK161621.1 | - | 2 | 122 | |||
BC011424.1 | - | 2 | 130 | 3674751 | ||
BY237508.1 | - | 2 | 192 | |||
JF720023.1 | - | 165 | ||||
JF720024.1 | - | 2 | 275 | |||
JF720025.1 | - | 2 | 193 | |||
M20473.1 | - | 2 | 137 |