Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Procr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011171.2 | NP_035301.2 | 3, 4 | 804 | ||
GenBank mRNA | AK136541.1 | - | 3, 4 | 665 | ||
AK143719.1 | - | 3, 4 | 906 | |||
AK167302.1 | - | 3, 4 | 744 | |||
BC028755.1 | - | 3 | 918 | 1054063 | ||
CT010175.1 | - | 3, 4 | 623 | |||
L39017.1 | - | 3, 4 | 678 | |||
X63748.2 | - | 3, 4 | 710 |