Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011187.1 | NP_035317.1 | 5 | 479 | ||
GenBank mRNA | AK002823.1 | - | 5 | 494 | ||
AK012613.1 | - | 5 | 480 | |||
AK013961.1 | - | 5 | 490 | |||
AK031913.1 | - | 5 | 495 | |||
AK075759.1 | - | 5 | 495 | |||
AK088276.1 | - | 5 | 490 | |||
AK088765.1 | - | 5 | 493 | |||
AK162002.1 | - | 5 | 498 | |||
AK168823.1 | - | 5 | 482 | |||
BC033424.1 | - | 5 | 476 | |||
BC049230.1 | - | 5 | 472 | |||
BC057662.1 | - | 5 | 480 | 2865594 | ||
D83585.1 | - | 5 | 479 | |||
Y10874.1 | - | 5 | 493 |