Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptafr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081211.1 | NP_001074680.1 | 2 | 475 | ||
GenBank mRNA | AK032547.1 | - | 2 | 319 |
Set Membership: |
![]() |