Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptger1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013641.2 | NP_038669.1 | 2 | 416 | ||
GenBank mRNA | AK053616.1 | - | 189 | |||
AK164746.1 | - | 224 | ||||
BC138739.1 | - | 2 | 378 | 8861761 | ||
BC138744.1 | - | 2 | 378 | 8861766 | ||
D16338.1 | - | 2 | 193 |
Set Membership: |
![]() |