Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019424.2 | NP_062297.1 | 12 | 1271 | ||
GenBank mRNA | AK031397.1 | - | 12 | 1285 | ||
AK053974.1 | - | 10 | 1103 | |||
AK154994.1 | - | 12 | 1460 | |||
AK171225.1 | - | 12 | 1247 | |||
BC046329.1 | - | 10 | 1092 | |||
BC058332.1 | - | 12 | 1254 | |||
BC089331.1 | - | 12 | 1254 | 6848298 | ||
U78315.1 | - | 11 | 1206 | |||
U97149.1 | - | 11 | 1220 |