Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081306.1 | NP_001074775.1 | 15 | 5447 | ||
GenBank mRNA | BC151071.1 | - | 15 | 5172 | 9087984 | |
BC157965.1 | - | 15 | 2625 | 9088134 |