Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008986.2 | NP_033012.1 | 1 | 610 | ||
GenBank mRNA | AF036249.1 | - | 1 | 565 | ||
AK009381.1 | - | 1 | 117 | |||
AK154132.1 | - | 1 | 547 | |||
AK169463.1 | - | 1 | 607 | |||
BC110698.1 | - | 1 | 592 | 30608521 | ||
BC116701.1 | - | 1 | 552 | 40104877 | ||
BC117527.1 | - | 1 | 552 | 40104876 |