Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ptrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008986.2 | NP_033012.1 | 2 | 969 | ||
GenBank mRNA | AF036249.1 | - | 2 | 924 | ||
BC110698.1 | - | 2 | 951 | 30608521 | ||
BC116701.1 | - | 2 | 911 | 40104877 | ||
BC117527.1 | - | 2 | 911 | 40104876 |