Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290633.1 | NP_001277562.1 | 13 | 1588 | ||
NM_178256.4 | NP_839987.2 | 13 | 1591 | |||
GenBank mRNA | AK147432.1 | - | 10 | 870 | ||
AY263368.1 | - | 13 | 1536 | |||
BC139012.1 | - | 13 | 1467 | 8862034 | ||
BC145500.1 | - | 13 | 1464 | 9054063 |