Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165934.1 | NP_001159406.1 | 5, 6 | 553 | ||
NM_011268.2 | NP_035398.2 | 5, 6 | 553 | |||
GenBank mRNA | AF011358.1 | - | 5, 6 | 438 | ||
AF125046.1 | - | 5, 6 | 509 | |||
AK085443.1 | - | 5, 6 | 479 | |||
AK138159.1 | - | 5, 6 | 463 | |||
AK164961.1 | - | 5, 6 | 507 | |||
BC129899.1 | - | 5, 6 | 419 | 40111531 |