Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011294.3 | NP_035424.1 | 3, 4 | 251 | ||
GenBank mRNA | AK048371.1 | - | 3, 4 | 251 | ||
AK049260.1 | - | 3, 4 | 251 | |||
AK132021.1 | - | 3, 4 | 250 | |||
AK146335.1 | - | 3, 4 | 250 | |||
AK151734.1 | - | 3, 4 | 251 | |||
AK154114.1 | - | 3, 4 | 250 | |||
BC010967.1 | - | 3, 4 | 249 | 4018261 | ||
J03750.1 | - | 3, 4 | 235 |