Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113248.2 | NP_001106719.1 | 5 | 615 | ||
NM_009188.4 | NP_033214.2 | 5 | 615 | |||
GenBank mRNA | AF038848.1 | - | 5 | 595 | ||
AK146847.1 | - | 5 | 598 | |||
AK159076.1 | - | 5 | 598 | |||
AK165592.1 | - | 5 | 600 | |||
AK169433.1 | - | 5 | 600 | |||
AK173005.1 | - | 5 | 584 | |||
BC020049.1 | - | 5 | 579 | |||
BC021160.1 | - | 5 | 598 | |||
BC051536.1 | - | 3 | 268 | |||
BC096367.1 | - | 5 | 583 | |||
BC129859.1 | - | 5 | 587 | |||
L38622.2 | - | 5 | 587 |