Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289725.1 | NP_001276654.1 | 3 | 541 | ||
NM_009193.2 | NP_033219.1 | 4 | 646 | |||
GenBank mRNA | AK131729.1 | - | 417 | |||
AK154053.1 | - | 4 | 393 | |||
AK162121.1 | - | 4 | 392 | |||
AK210098.1 | - | 3 | 185 | |||
BC025113.1 | - | 4 | 381 | |||
U75680.1 | - | 4 | 378 |