Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005045.2 | - | 2 | 207 | ||
AK011564.1 | - | 2 | 163 | |||
AK014311.2 | - | 2 | 204 | |||
AK077425.1 | - | 2 | 209 | |||
AK133115.1 | - | 2 | 208 | |||
AK146729.1 | - | 2 | 208 | |||
AK148013.1 | - | 2 | 234 | |||
BC049128.1 | - | 2 | 173 | |||
BC131994.1 | - | 2 | 100 | 40130271 | ||
BC132020.1 | - | 2 | 100 | 40130297 | ||
BY297843.1 | - | 2 | 257 |