Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011439.2 | NP_035569.2 | 4 | 710 | ||
GenBank mRNA | AB006329.1 | - | 3 | 366 | ||
AJ000740.1 | - | 3 | 320 | |||
BC007130.1 | - | 4 | 532 | 3493465 |