Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sall3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178280.3 | NP_840064.2 | 2 | 1615 | ||
GenBank mRNA | BC072631.1 | - | 2 | 1559 | ||
BC118974.1 | - | 1 | 1893 | |||
BC128071.1 | - | 1 | 1892 | |||
X97581.1 | - | 1 | 1487 |