Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177667.1 | NP_001171138.1 | 2 | 422 | ||
NM_001177668.1 | NP_001171139.1 | 2 | 422 | |||
GenBank mRNA | AK037683.1 | - | 2 | 306 | ||
AK132224.1 | - | 2 | 306 | |||
AK141149.1 | - | 2 | 303 | |||
AK147317.1 | - | 2 | 306 | |||
BC150941.1 | - | 2 | 258 | 9087852 | ||
BC158015.1 | - | 2 | 258 | 9007321 | ||
BC172095.1 | - | 2 | 258 | 9054381 | ||
CJ054332.1 | - | 2 | 423 |