Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164493.1 | NP_001157965.1 | 13, 14 | 3186 | ||
GenBank mRNA | AK039195.1 | - | 2, 3 | 287 | ||
AK122572.1 | - | 13, 14 | 2839 | |||
BC065168.1 | - | 13, 14 | 3186 | |||
BC138283.1 | - | 12, 13 | 2542 | 8861302 | ||
BC145230.1 | - | 11, 12 | 2359 | |||
BC145748.1 | - | 12, 13 | 2542 | 40131051 |