Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011484.2 | NP_035614.1 | 12 | 1259 | ||
GenBank mRNA | AK035924.1 | - | 12 | 1182 | ||
AK144987.1 | - | 12 | 1276 | |||
AK147832.1 | - | 12 | 1169 | |||
AK157767.1 | - | 12 | 1206 | |||
AK162431.1 | - | 12 | 1206 | |||
AK162973.1 | - | 7 | 444 | |||
AK163720.1 | - | 12 | 1209 | |||
AK163922.1 | - | 12 | 1196 | |||
AK164191.1 | - | 4 | 191 | |||
AK169170.1 | - | 12 | 1266 | |||
BC044666.1 | - | 12 | 1259 | 4950569 | ||
BC055326.1 | - | 12 | 1205 | 6416552 | ||
U43900.1 | - | 12 | 1116 |