Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009285.3 | NP_033311.3 | 2 | 311 | ||
GenBank mRNA | AF099098.1 | - | 2 | 363 | ||
AK134260.1 | - | 2 | 312 | |||
AK142220.1 | - | 2 | 210 | |||
AK149293.1 | - | 2 | 194 | |||
AK149351.1 | - | 2 | 194 | |||
AK161960.1 | - | 2 | 193 | |||
BC021425.1 | - | 2 | 332 | 5010981 |