Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145947.2 | NP_666059.2 | 9, 10 | 1268 | ||
GenBank mRNA | AF345194.1 | - | 8, 9 | 1170 | ||
AK054327.1 | - | 9, 10 | 1269 | |||
BC026928.1 | - | 9, 10 | 1247 | 4020760 |