Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stra13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016665.2 | NP_057874.2 | 2, 3 | 117 | ||
GenBank mRNA | AK080500.1 | - | 2, 3 | 89 | ||
AK170126.1 | - | 2, 3 | 89 | |||
BC052335.1 | - | 3, 4 | 367 | |||
CA320983.1 | - | 1, 2 | 56 | |||
CJ236047.1 | - | 2, 3 | 118 | |||
U95003.1 | - | 3, 4 | 181 | |||
U95004.1 | - | 2, 3 | 82 | |||
U95005.1 | - | 3, 4 | 355 |