Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009292.1 | NP_033318.1 | 7, 8 | 1093 | ||
GenBank mRNA | AK006887.1 | - | 8, 9 | 1434 | ||
BC100746.1 | - | 6, 7 | 841 | |||
BC103590.1 | - | 7, 8 | 1039 | 40045824 | ||
BC103591.1 | - | 7, 8 | 1039 | 40045823 | ||
BC103603.1 | - | 7, 8 | 1036 | |||
Z75287.1 | - | 7, 8 | 1093 |