Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025307.1 | NP_001020478.1 | 1, 2 | 721 | ||
NM_001286543.1 | NP_001273472.1 | 1, 2 | 721 | |||
NM_011502.3 | NP_035632.1 | 1, 2 | 721 | |||
NM_152220.2 | NP_689344.1 | 1, 2 | 721 | |||
GenBank mRNA | AK038415.1 | - | 1, 2 | 228 | ||
AK133830.1 | - | 1, 2 | 244 | |||
AK170105.1 | - | 1, 2 | 130 | |||
AK171135.1 | - | 1, 2 | 264 | |||
BC024844.1 | - | 1, 2 | 129 | 5357204 | ||
BC056949.1 | - | 1, 2 | 721 | |||
D29797.1 | - | 1, 2 | 142 | |||
D29798.1 | - | 1, 2 | 140 | |||
D29799.1 | - | 1, 2 | 209 | |||
D29800.1 | - | 1, 2 | 53 | |||
D38375.1 | - | 1, 2 | 39 |