Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011504.1 | NP_035634.1 | 18 | 1587 | ||
GenBank mRNA | AK138609.1 | - | 18 | 1619 | ||
AK138769.1 | - | 6 | 870 | |||
AK151466.1 | - | 18 | 1582 | |||
AK152261.1 | - | 18 | 1576 | |||
AK155133.1 | - | 18 | 1585 | |||
AK169799.1 | - | 16 | 1421 | |||
BC062901.1 | - | 18 | 1596 | 6810254 | ||
BC094055.1 | - | 18 | 1595 | |||
D30798.1 | - | 18 | 1587 | |||
U19521.1 | - | 18 | 1712 |