Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Frmd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177843.1 | NP_001171314.1 | 20, 21 | 2629 | ||
NM_001177844.1 | NP_001171315.1 | 9, 10 | 1757 | |||
NM_172475.3 | NP_766063.3 | 20, 21 | 2259 | |||
GenBank mRNA | AB093292.1 | - | 6, 7 | 953 | ||
AK082365.1 | - | 20, 21 | 2205 | |||
AK148414.1 | - | 9, 10 | 1756 | |||
AK171532.1 | - | 9, 10 | 1685 | |||
BC058672.1 | - | 20, 21 | 2250 | 6830456 | ||
BC082808.1 | - | 20, 21 | 2636 |