Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172378.2 | NP_759010.1 | 9 | 1563 | ||
GenBank mRNA | AB520670.1 | - | 8 | 1169 | ||
AK030222.1 | - | 9 | 1563 | |||
BC026439.1 | - | 8 | 1206 | 4239636 |