Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081080.1 | NP_001074549.1 | 3 | 1913 | ||
GenBank mRNA | AK135686.1 | - | 1 | 665 | ||
BC011226.1 | - | 3 | 1908 | |||
BC019710.1 | - | 1 | 393 | |||
BC032205.1 | - | 3 | 1927 | |||
BC038053.1 | - | 1 | 323 | |||
BC137913.1 | - | 3 | 1919 | 8860932 |