Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubtf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004961.1 | - | 9 | 1056 | ||
AK038139.1 | - | 5 | 2186 | |||
AK128947.1 | - | 9 | 1083 | |||
AK158861.1 | - | 9 | 1055 | |||
AK158935.1 | - | 9 | 1014 | |||
BC004676.1 | - | 10 | 1300 | |||
CK392577.1 | - | 5 | 487 | |||
X60831.1 | - | 9 | 1034 |