Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnptg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172529.3 | NP_766117.2 | 2, 3 | 119 | ||
GenBank mRNA | AK078230.1 | - | 127 | |||
AY465529.1 | - | 2, 3 | 127 | |||
BC055872.1 | - | 2, 3 | 119 | 4925332 |