Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290278.1 | NP_001277207.1 | 6 | 746 | ||
NM_001290279.1 | NP_001277208.1 | 6 | 746 | |||
NM_001290280.1 | NP_001277209.1 | 6 | 746 | |||
NM_001290282.1 | NP_001277211.1 | 7 | 860 | |||
NM_144814.3 | NP_659063.3 | 6 | 746 | |||
GenBank mRNA | AK040839.1 | - | 6 | 733 | ||
AK050287.1 | - | 7 | 851 | |||
AK135612.1 | - | 6 | 719 | |||
AK155440.1 | - | 6 | 718 | |||
AK193623.1 | - | 2 | 180 | |||
BC020065.1 | - | 6 | 633 | |||
BC057141.1 | - | 6 | 493 | 5151580 | ||
BC138938.1 | - | 6 | 466 | 8861960 | ||
BC138939.1 | - | 6 | 466 | 8861961 | ||
BC145469.1 | - | 6 | 466 |