Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maneal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007573.2 | NP_001007574.1 | 4 | 1188 | ||
GenBank mRNA | AK137071.1 | - | 1 | 218 | ||
BC065047.1 | - | 1207 | 5694626 |