Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271356.1 | NP_001258285.1 | 9 | 1631 | ||
NM_001271357.1 | NP_001258286.1 | 9 | 1374 | |||
NM_146018.2 | NP_666130.1 | 9 | 1671 | |||
GenBank mRNA | AK039106.1 | - | 9 | 1458 | ||
AK080933.1 | - | 9 | 1456 | |||
AK136071.1 | - | 9 | 1362 | |||
AK148216.1 | - | 9 | 1425 | |||
AK154040.1 | - | 9 | 1653 | |||
AK161460.1 | - | 9 | 1403 | |||
AK172245.1 | - | 9 | 1429 | |||
AK177194.1 | - | 3 | 360 | |||
AK179995.1 | - | 2 | 122 | |||
AK180220.1 | - | 2 | 122 | |||
AK194459.1 | - | 2 | 122 | |||
AK213109.1 | - | 2 | 122 | |||
BC025820.1 | - | 9 | 1405 | 5099493 |