Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172050.2 | NP_742047.1 | 2 | 177 | ||
GenBank mRNA | AF437879.1 | - | 2 | 201 | ||
AY457048.1 | - | 2 | 163 | |||
BC034097.1 | - | 2 | 202 | 4487018 | ||
BC039971.1 | - | 2 | 213 | 4487018 |