Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Terf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009352.3 | NP_033378.1 | 4, 5 | 626 | ||
GenBank mRNA | AB109563.1 | - | 4, 5 | 606 | ||
AK021235.1 | - | 4, 5 | 616 | |||
AK036615.1 | - | 4, 5 | 629 | |||
BC034866.1 | - | 4, 5 | 614 | 3470277 | ||
U65586.1 | - | 4, 5 | 606 |