Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis18bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172578.2 | NP_766166.2 | 8 | 2267 | ||
GenBank mRNA | AK032698.1 | - | 6 | 1299 | ||
AK035117.1 | - | 8 | 2266 | |||
AK220223.1 | - | 7 | 1533 | |||
BC021832.1 | - | 2, 3 | 810 | |||
BC052175.1 | - | 8 | 2267 | 6394068 |