Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173757.3 | NP_776118.2 | 8 | 680 | ||
GenBank mRNA | AK076008.1 | - | 8 | 679 | ||
AK087686.1 | - | 8 | 677 | |||
AK172922.1 | - | 1 | 2099 | |||
BC049149.1 | - | 8 | 607 | 6309524 |
Gene Symbol: 6430562O15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032482.1 | - | 2724 | |||
AK044404.1 | - | 2782 |