Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccno | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081062.1 | NP_001074531.1 | 3 | 856 | ||
GenBank mRNA | AK086507.1 | - | 3 | 831 | ||
BC147759.1 | - | 3 | 832 | 9087484 | ||
BC147766.1 | - | 3 | 832 | 9087491 |