Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027906.1 | NP_082182.1 | 24 | 2846 | ||
NM_173758.3 | NP_776119.2 | 24 | 2846 | |||
GenBank mRNA | AK033642.1 | - | 24 | 2794 | ||
AK129164.1 | - | 8 | 765 | |||
AK154108.1 | - | 24 | 2830 | |||
BC019143.1 | - | 24 | 2778 | 5042800 |