Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009393.2 | NP_033419.1 | 4 | 268 | ||
GenBank mRNA | BC061172.1 | - | 4 | 262 | 30300744 | |
M29793.1 | - | 4 | 268 |