Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020557.4 | NP_065582.3 | 5 | 1467 | ||
GenBank mRNA | AK004595.1 | - | 5 | 1302 | ||
AK150512.1 | - | 5 | 1292 | |||
AK150543.1 | - | 5 | 1294 | |||
AK150725.1 | - | 5 | 1292 | |||
AK150885.1 | - | 5 | 1294 | |||
AK153429.1 | - | 5 | 1296 | |||
AK153469.1 | - | 5 | 1299 | |||
BC027329.1 | - | 5 | 1366 | 3982036 | ||
BC048910.1 | - | 5 | 1479 | |||
BC057565.1 | - | 5 | 1280 | 6401867 | ||
L32973.1 | - | 4 | 1553 |