Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyrobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011662.2 | NP_035792.1 | 1 | 72 | ||
GenBank mRNA | AF024637.1 | - | 1 | 21 | ||
AF077829.1 | - | 1 | 21 | |||
AK154480.1 | - | 1 | 71 | |||
BC056450.1 | - | 1 | 72 | 6814478 |