Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpysl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136086.2 | NP_001129558.1 | 10 | 1157 | ||
NM_001291455.1 | NP_001278384.1 | 10 | 1407 | |||
NM_009468.5 | NP_033494.1 | 10 | 1338 | |||
GenBank mRNA | AK077000.1 | - | 10 | 1148 | ||
AK133040.1 | - | 10 | 1157 | |||
AK149024.1 | - | 10 | 1175 | |||
AK161509.1 | - | 10 | 1079 | |||
AK171653.1 | - | 3 | 238 | |||
BC023003.1 | - | 10 | 1022 | 5367381 | ||
X87817.1 | - | 10 | 1298 |