Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011681.2 | NP_035811.1 | 2 | 133 | ||
GenBank mRNA | AK145844.1 | - | 2 | 98 | ||
BC027518.1 | - | 2 | 90 | 1434396 | ||
CB595042.1 | - | 2 | 137 | |||
L04503.1 | - | 2 | 159 | |||
X67702.1 | - | 2 | 140 |
Gene Symbol: Bpifb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC089529.1 | - | 895 |